

α,α,α,α',α',α'-Hexafluoro-2,4-xylidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367-71-5
:
Formula:C₈H₅F₆N
MDL Number:MFCD00236209
:229.12 g/mol
Boiling Pt:50 °C/0.4mmHg
UN:2810
ADR:6.1,III
:GHS07
:GHS08
Storage Temperature:Ambient