Order Entry
Export
ContactUsLinkComponent
2,4-Difluorophenylglyoxal hydrate 95%
2,4-Difluorophenylglyoxal hydrate 95%
Catalog # APOSPC2005-5G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Difluorophenylglyoxal hydrate 95%
Catalog # APOSPC2005-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :188.13 g/mol
Storage Temperature:Ambient
CAS Number:79784-36-4
Formula:C₈H₆F₂O₃
MDL Number:MFCD04038287