4-Chloro-N-(2,5-difluorophenyl)benzenesulphonamide
Catalog # APOSPC200495-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:303.72 g/mol
Storage Temperature:Ambient
CAS Number:290331-05-4
Formula:C₁₂H₈ClF₂NO₂S
MDL Number:MFCD05136552