Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
Supplier:
Apollo Scientific
Certificates
Expand image
4,6-Dichloro-2-(trifluoromethyl)pyrimidine
Supplier:
Apollo Scientific
About this item
CAS Number:
705-24-8
:
Formula:
C₅HCl₂F₃N₂
MDL Number:
MFCD08436597
:
216.98 g/mol
Storage Temperature:
Ambient