Order Entry
Export
ContactUsLinkComponent
4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
Catalog # APOSPC200357-500MG
Supplier:  Apollo Scientific
CAS Number:  
4,4-Diethoxy-1,1,1-trifluorobut-3-en-2-one
Catalog # APOSPC200357-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :212.17 g/mol
Boiling Pt:110…122 °C/13mmHg
Storage Temperature:Ambient
CAS Number:40657-29-2
Formula:C₈H₁₁F₃O₃
MDL Number:MFCD08273801