Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichloro-5-(trifluoromethyl)-4-pyridinamine
2,3-Dichloro-5-(trifluoromethyl)-4-pyridinamine
2,3-Dichloro-5-(trifluoromethyl)-4-pyridinamine
2,3-Dichloro-5-(trifluoromethyl)-4-pyridinamine
Supplier:
Apollo Scientific
Certificates
Expand image
2,3-Dichloro-5-(trifluoromethyl)-4-pyridinamine
Supplier:
Apollo Scientific
About this item
CAS Number:
431942-80-2
:
Formula:
C₆H₃Cl₂F₃N₂
MDL Number:
MFCD14581629
:
231.01 g/mol
Melting Pt:
130…131 °C
Storage Temperature:
Ambient