
5-Fluoroimidazo[1,2-a]pyridine-2-carbaldehyde
Catalog # APOSPC200220-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:164.14 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:878197-67-2
Formula:C₈H₅FN₂O
MDL Number:MFCD11656369