Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4,6-bis(trifluoromethyl)nicotinamide
2-Amino-4,6-bis(trifluoromethyl)nicotinamide
2-Amino-4,6-bis(trifluoromethyl)nicotinamide
2-Amino-4,6-bis(trifluoromethyl)nicotinamide
Supplier:
Apollo Scientific
Certificates
Expand image
2-Amino-4,6-bis(trifluoromethyl)nicotinamide
Supplier:
Apollo Scientific
About this item
CAS Number:
89990-38-5
:
Formula:
C₈H₅F₆N₃O
MDL Number:
MFCD13193070
:
273.14 g/mol
Melting Pt:
188…190 °C
Storage Temperature:
Ambient