Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
:
Apollo Scientific
8-Iodo-6-(trifluoromethyl)imidazo[1,2-a]pyridine
:
Apollo Scientific
:
1048913-78-5
:
:
C₈H₄F₃IN₂
:
MFCD11100236
:
312.03 g/mol
:
114…116 °C
:
GHS07
:
Refrigerator