Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,8-Dichloro-6-(trifluoromethyl)quinoline
4,8-Dichloro-6-(trifluoromethyl)quinoline 95+%
4,8-Dichloro-6-(trifluoromethyl)quinoline 95+%
Irritant
4,8-Dichloro-6-(trifluoromethyl)quinoline 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,8-Dichloro-6-(trifluoromethyl)quinoline 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1065093-22-2
:
Formula:
C₁₀H₄Cl₂F₃N
Melting Pt:
124 °C
:
GHS07
Storage Temperature:
Ambient