Order Entry
Export
ContactUsLinkComponent
1-(3,5-Difluorophenyl)ethanol 97%
Irritant
1-(3,5-Difluorophenyl)ethanol 97%
Catalog # APOSPC2001-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(3,5-Difluorophenyl)ethanol 97%
Catalog # APOSPC2001-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :158.15 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:872181-59-4
Formula:C₈H₈F₂O
MDL Number:MFCD04038285