
Ethyl-6-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:367500-93-4
:
Formula:C₁₀H₉FN₂O₂
MDL Number:MFCD06659035
:208.1900 g/mol
Melting Pt:146…147 °C
:GHS07
Storage Temperature:Ambient