N-[3-(Trifluoromethyl)phenyl]-1,3,5-triazine-2,4-diamine
Catalog # APOSPC200029-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:255.2 g/mol
Storage Temperature:Ambient
CAS Number:3832-69-7
Formula:C₁₀H₈F₃N₅
MDL Number:MFCD00179648