2-Chloro-5-fluoroaniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:452-83-5
:
Formula:C₆H₅ClFN
MDL Number:MFCD00084852
:145.56 g/mol
Boiling Pt:211 °C
UN:2810
ADR:6.1,III
:GHS06
:GHS07
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: