

2-Chloro-3,5-difluorophenol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:206986-81-4
:
Formula:C₆H₃ClF₂O
MDL Number:MFCD00075400
:164.54 g/mol
Boiling Pt:76 °C/37mmHg
Melting Pt:45…46 °C
Flash Pt:66 °C
Storage Temperature:Ambient
UN:1325
ADR:4.1,II
:GHS02
:GHS07