
2-Chloro-1,4-dibromo-1,1,2-trifluorobutane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:378-13-2
:
Formula:C₄H₄Br₂ClF₃
MDL Number:MFCD00017985
:304.33 g/mol
Boiling Pt:70…75 °C/26mmHg
Density:2.035 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient