Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,6-difluoroanisole
4-Bromo-2,6-difluoroanisole 98%
4-Bromo-2,6-difluoroanisole 98%
:
Apollo Scientific
4-Bromo-2,6-difluoroanisole 98%
:
Apollo Scientific
:
104197-14-0
:
:
C₇H₅BrF₂O
:
MFCD01631351
:
223.02 g/mol
:
100 °C/15mmHg
:
1.65 g/cm³ (25 °C)
:
GHS07
:
Ambient