
3,4-Dichloro-6-(trifluoromethyl)toluene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74483-51-5
:
Formula:C₈H₅Cl₂F₃
MDL Number:MFCD06246858
:229.03 g/mol
Boiling Pt:97…100 °C/23mmHg
Flash Pt:88 °C
:GHS07
Storage Temperature:Ambient