Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-2,4-difluorobenzonitrile
3-Chloro-2,4-difluorobenzonitrile 98%
3-Chloro-2,4-difluorobenzonitrile 98%
APOSPC1503-1G
:
Apollo Scientific
:
3-Chloro-2,4-difluorobenzonitrile 98%
APOSPC1503-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
173.5500 g/mol
:
Ambient
:
887267-38-1
:
C7H2ClF2N
:
MFCD06658267