
2-Bromo-1,4-difluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:399-94-0
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00000345
:192.99 g/mol
Boiling Pt:58…59 °C/20mmHg
Melting Pt:–31 °C
Density:1.708 g/cm³ (25 °C)
Flash Pt:65 °C
EINECS:206-920-0
:GHS07
Storage Temperature:Ambient