

4-Bromo-1,2-difluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:348-61-8
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00000304
:192.99 g/mol
Boiling Pt:150…151 °C
Melting Pt:–4 °C
Density:1.707 g/cm³ (25 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
EINECS:206-481-5
UN:1993
ADR:3,III
:GHS02
:GHS07