
1-Bromo-2,3-difluorobenzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:38573-88-5
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00061136
:192.99 g/mol
Boiling Pt:234 °C/765mmHg
Density:1.724 g/cm³ (25 °C)
Flash Pt:61 °C
:GHS07
Storage Temperature:Ambient