0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 98%
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 98%
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Bromo-3-chloro-3,4,4-trifluorobut-1-ene 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
374-25-4
:
Formula:
C₄H₃BrClF₃
MDL Number:
MFCD00039225
:
223.43 g/mol
Boiling Pt:
100 °C
Storage Temperature:
Ambient