2,3-Difluorophenetole
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 1-Ethoxy-2,3-difluorobenzene, 2,3-Difluoroethoxybenzene
- CAS Number:121219-07-6
- Formula:C₈H₈F₂O
- MDL Number:MFCD07368737
- Molecular Weight:158.15 g/mol
- Boiling Pt:178 °C
- Density:1.18 g/cm³ (25 °C)
- Flash Pt:70.5 °C
- Storage Temperature:Ambient
- EINECS:441-000-4
3 Options Available Below
Product Details & Documents
Synonyms: 1-Ethoxy-2,3-difluorobenzene, 2,3-Difluoroethoxybenzene
- CAS Number:121219-07-6
- Formula:C₈H₈F₂O
- MDL Number:MFCD07368737
- Molecular Weight:158.15 g/mol
- Boiling Pt:178 °C
- Density:1.18 g/cm³ (25 °C)
- Flash Pt:70.5 °C
- Storage Temperature:Ambient
- EINECS:441-000-4



