Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Fluoro-1,3-phenylenediamine
4-Fluoro-1,3-phenylenediamine 98%
4-Fluoro-1,3-phenylenediamine 98%
4-Fluoro-1,3-phenylenediamine 98%
APOSPC1338-5G
:
Apollo Scientific
:
4-Fluoro-1,3-phenylenediamine 98%
APOSPC1338-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
126.13 g/mol
:
Ambient
:
6264-67-1
:
C₆H₇FN₂
:
MFCD00077494