2,2-Difluoropropanol
Bottle, 1 g
Catalog # APOSPC1247-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2,2-Difluoropropan-1-ol
- CAS Number:33420-52-9
- Formula:C₃H₆F₂O
- MDL Number:MFCD07782150
- Molecular Weight:96.08 g/mol
- Boiling Pt:97…99 °C
- Density:1.1642 g/cm³ (25 °C)
- Flash Pt:31.1 °C
- Storage Temperature:Ambient
- UN:1987
- ADR:3,III
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



