Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
:
Apollo Scientific
1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene 97%
:
Apollo Scientific
:
1992-15-0
:
:
C₁₂H₆F₁₂O₂
:
MFCD00042091
:
410.16 g/mol
:
99 °C/20mmHg
:
87 °C
:
1.659 g/cm³ (25 °C)
:
108 °C
:
Ambient
:
3077
:
9+EHS,III
:
GHS07
:
GHS09