2,2-Bis(3,4-dimethylphenyl)hexafluoropropane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65294-20-4
:
Formula:C₁₉H₁₈F₆
MDL Number:MFCD00042167
Boiling Pt:110…120 °C/2mmHg
Melting Pt:76…78 °C
:360.34 g/mol
Storage Temperature:Ambient