Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Bis(tert-butoxy)tetrafluorobenzene
1,4-Bis(tert-butoxy)tetrafluorobenzene, Technical Grade
1,4-Bis(tert-butoxy)tetrafluorobenzene, Technical Grade
1,4-Bis(tert-butoxy)tetrafluorobenzene, Technical Grade
Supplier:
Apollo Scientific
Certificates
Expand image
1,4-Bis(tert-butoxy)tetrafluorobenzene, Technical Grade
Supplier:
Apollo Scientific
About this item
CAS Number:
121088-09-3
:
Formula:
C₁₄H₁₈F₄O₂
MDL Number:
MFCD00069107
:
294.29 g/mol
Melting Pt:
55…56 °C
Storage Temperature:
Ambient