

4-(3,5-Bis(trifluormethyl)phenyl)aniline
Catalog # APOSPC11219-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS05
:GHS06
CAS Number:444143-45-7
Formula:C₁₄H₉F₆N
MDL Number:MFCD05154989