
2,4-Dichloro-5-fluorobenzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:128593-93-1
:
Formula:C₇H₂Cl₂FN
MDL Number:MFCD03840469
:190.00 g/mol
Melting Pt:44…47 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07