Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Difluoro-2-hydroxybenzonitrile
4,5-Difluoro-2-hydroxybenzonitrile
4,5-Difluoro-2-hydroxybenzonitrile
APOSPC1116-250MG
:
Apollo Scientific
:
4,5-Difluoro-2-hydroxybenzonitrile
APOSPC1116-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
155.10 g/mol
:
161…163 °C
:
GHS07
:
Ambient
:
186590-36-3
:
C₇H₃F₂NO
:
MFCD08275287