Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Difluoro-3-nitrobenzene
1,2-Difluoro-3-nitrobenzene 97%
1,2-Difluoro-3-nitrobenzene 97%
:
Apollo Scientific
1,2-Difluoro-3-nitrobenzene 97%
:
Apollo Scientific
:
6921-22-8
:
:
C₆H₃F₂NO₂
:
MFCD00456802
:
159.09 g/mol
:
90 °C
:
GHS07
:
Ambient