Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluoropicolinic acid
3,5-Difluoropicolinic acid
3,5-Difluoropicolinic acid
Irritant
3,5-Difluoropicolinic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Difluoropicolinic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
745784-04-7
:
Formula:
C₆H₃F₂NO₂
MDL Number:
MFCD05664124
Melting Pt:
202…204 °C
:
159.09 g/mol
:
GHS07
Storage Temperature:
Ambient