Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-2-fluoropyridine
3,5-Dichloro-2-fluoropyridine 98%
3,5-Dichloro-2-fluoropyridine 98%
Irritant
3,5-Dichloro-2-fluoropyridine 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dichloro-2-fluoropyridine 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
823-56-3
:
Formula:
C₅H₂Cl₂FN
MDL Number:
MFCD04039349
Melting Pt:
41…44 °C
:
165.98 g/mol
:
GHS07
Storage Temperature:
Ambient