Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1-Difluoro-2-iodoethane
1,1-Difluoro-2-iodoethane 97%
1,1-Difluoro-2-iodoethane 97%
Irritant
1,1-Difluoro-2-iodoethane 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,1-Difluoro-2-iodoethane 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
598-39-0
:
Formula:
C₂H₃F₂I
Boiling Pt:
86.8…87 °C/740mmHg
Density:
2.18 g/cm³ (25 °C)
:
GHS07
Storage Temperature:
Refrigerator