Order Entry
Export
ContactUsLinkComponent
1-Chloro-1,2-dibromo-2-fluoropropane 97%
1-Chloro-1,2-dibromo-2-fluoropropane 97%
Catalog # APOSPC1032-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Chloro-1,2-dibromo-2-fluoropropane 97%
Catalog # APOSPC1032-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:885276-04-0
Formula:C₃H₄Br₂ClF