6-(2-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:439095-02-0
:
Formula:C₁₂H₇FN₂OS
MDL Number:MFCD02091516
:246.26 g/mol
Melting Pt:147…148 °C
Storage Temperature:Ambient