Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Amino-2,4-difluorophenol
5-Amino-2,4-difluorophenol
5-Amino-2,4-difluorophenol
Irritant
5-Amino-2,4-difluorophenol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Amino-2,4-difluorophenol
Supplier:
Apollo Scientific
About this item
CAS Number:
113512-71-3
:
Formula:
C₆H₅F₂NO
MDL Number:
MFCD04973718
:
145.11 g/mol
Melting Pt:
177 °C (dec.)
:
GHS07
Storage Temperature:
Refrigerator