2-(3,4-Difluorophenoxy)benzaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:289717-89-1
:
Formula:C₁₃H₈F₂O₂
MDL Number:MFCD01568793
Boiling Pt:110…112 °C/0.2mmHg
Melting Pt:61…64 °C
Storage Temperature:Ambient
:234.1982 g/mol