1,1-Difluoroacetone 97%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:431-05-0
- Formula:C₃H₄F₂O
- Boiling Pt:46.5…46.7 °C/757mmHg
- Density:1.164 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:1224
- ADR:3,II
3 Options Available Below
Product Details & Documents
- CAS Number:431-05-0
- Formula:C₃H₄F₂O
- Boiling Pt:46.5…46.7 °C/757mmHg
- Density:1.164 g/cm³ (25 °C)
- Storage Temperature:Refrigerator
- UN:1224
- ADR:3,II



