
7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate 95+%
Catalog # APOSPC0963-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:254.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:849924-90-9
Formula:C₁₀H₇FN₂O₃S
MDL Number:MFCD08445326