Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,5-difluorobenzenesulphonyl chloride
2-Bromo-4,5-difluorobenzenesulphonyl chloride
2-Bromo-4,5-difluorobenzenesulphonyl chloride
:
Apollo Scientific
2-Bromo-4,5-difluorobenzenesulphonyl chloride
:
Apollo Scientific
:
874784-11-9
:
:
C₆H₂BrClF₂O₂S
:
MFCD04035650
:
291.5 g/mol
:
Ambient
:
3261
:
8,III
:
GHS05