Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,5-difluorobenzoyl chloride
4-Chloro-2,5-difluorobenzoyl chloride 97%
4-Chloro-2,5-difluorobenzoyl chloride 97%
Corrosive
Irritant
4-Chloro-2,5-difluorobenzoyl chloride 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
Irritant
4-Chloro-2,5-difluorobenzoyl chloride 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
132794-08-2
:
Formula:
C₇H₂Cl₂F₂O
MDL Number:
MFCD00236600
:
210.99 g/mol
UN:
3265
ADR:
8,III
:
GHS05
:
GHS07
Storage Temperature:
Ambient