Order Entry
Export
ContactUsLinkComponent
2,2,3,3-Tetrafluoro-1,4-benzodioxane
Irritant
2,2,3,3-Tetrafluoro-1,4-benzodioxane
Catalog # APOSPC0789-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2,3,3-Tetrafluoro-1,4-benzodioxane
Catalog # APOSPC0789-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:94767-47-2
Formula:C₈H₄F₄O₂
MDL Number:MFCD00236308