

2,6-Difluoroanisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:437-82-1
:
Formula:C₇H₆F₂O
MDL Number:MFCD00142846
:144.12 g/mol
Boiling Pt:70…72 °C/56mmHg
Density:1.221 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07