
α,α,α',α'-Tetrafluoro-p-xylene 99%
Catalog # APOSPC0502-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:178.13 g/mol
Boiling Pt:83…84 °C/30mmHg
Density:1.312 g/cm³ (25 °C)
Flash Pt:63 °C
:GHS07
Storage Temperature:Ambient
CAS Number:369-54-0
Formula:C₈H₆F₄
MDL Number:MFCD01320708