
4-Bromo-3,5-difluorobenzotrifluoride 95%
Catalog # APOSPC0321-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260.99 g/mol
Density:1.74 g/cm³ (25 °C)
Flash Pt:78 °C
:GHS07
Storage Temperature:Ambient
CAS Number:499238-36-7
Formula:C₇H₂BrF₅
MDL Number:MFCD03093034