Order Entry
Export
ContactUsLinkComponent
2,6-Difluoro-3-methylbenzamide 97%
2,6-Difluoro-3-methylbenzamide 97%
Catalog # APOSPC0244-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Difluoro-3-methylbenzamide 97%
Catalog # APOSPC0244-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:261763-36-4
Formula:C₈H₇F₂NO
MDL Number:MFCD01631408