Order Entry
Export
ContactUsLinkComponent
2,3-Difluoro-4-methylbenzamide 98%
2,3-Difluoro-4-methylbenzamide 98%
Catalog # APOSPC0241-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Difluoro-4-methylbenzamide 98%
Catalog # APOSPC0241-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.15 g/mol
Storage Temperature:Ambient
CAS Number:261763-35-3
Formula:C₈H₇F₂NO
MDL Number:MFCD01631633